Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B8750

Sigma-Aldrich

1-Benzylindole-3-carboxaldehyde

Synonym(s):

1-Benzyl-3-formylindole, NSC 95434

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13NO
CAS Number:
Molecular Weight:
235.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

O=Cc1cn(Cc2ccccc2)c3ccccc13

InChI

1S/C16H13NO/c18-12-14-11-17(10-13-6-2-1-3-7-13)16-9-5-4-8-15(14)16/h1-9,11-12H,10H2

InChI key

OXCITQLDOUGVRZ-UHFFFAOYSA-N

Application

  • reactant in preparation of inhibitor of the C-terminal domain of RNA polymerase II[1]
  • reactant in Nazarov type electrocyclization[2]
  • reactant in preparation inhibitors of Bcl-2 family of proteins[3]
  • reactant for Mannich type coupling with aldehydes and secondary amines[4]

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ritesh Singh et al.
Organic & biomolecular chemistry, 9(13), 4782-4790 (2011-02-25)
An easy, efficient and concise approach to tetrahydrofluorene [6,5,6]ABC tricyclic core embedded new polycycles has been achieved under relatively mild and catalytic Nazarov type electrocyclization conditions, using 2 mol% of Sc(OTf)(3) in anhydrous DCM (dichloromethane) at room temperature, with high
Bingcheng Zhou et al.
ChemMedChem, 6(5), 904-921 (2011-04-27)
A class of compounds with a common thiazolo[3,2-a]pyrimidinone motif has been developed as general inhibitors of Bcl-2 family proteins. The lead compound was originally identified in a random screening of a small compound library using a fluorescence polarization-based competitive binding
Atlan, V.; et al.
Chemical Communications (Cambridge, England), 7, 918-918 (2003)
Shuhong Wu et al.
Journal of medicinal chemistry, 54(8), 2668-2679 (2011-03-30)
To optimize the antitumor activity of oncrasin-1, a small molecule RNA polymerase II inhibitor, we evaluated 69 oncrasin-1 analogues for their cytotoxic activity against normal human epithelial cells and K-Ras mutant tumor cells. About 40 of those compounds were as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service