Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

Skip To

B39593

1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane

97%

Synonym(s):

4,4′-DDD, TDE

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
(ClC6H4)2CHCHCl2
CAS Number:
Molecular Weight:
320.04
EC Number:
200-783-0
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1914072
MDL number:

assay

97%

mp

109-111 °C

SMILES string

ClC(Cl)C(c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C14H10Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8,13-14H

InChI key

AHJKRLASYNVKDZ-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service