Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B24154

Sigma-Aldrich

N-Benzylidenemethylamine

97%

Synonym(s):

N-Methyl-α-tolueneimine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=NCH3
CAS Number:
Molecular Weight:
119.16
Beilstein/REAXYS Number:
878278
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

liquid

refractive index

n20/D 1.552 (lit.)

bp

75-80 °C/18 mmHg (lit.)

density

0.967 g/mL at 25 °C (lit.)

SMILES string

C\N=C\c1ccccc1

InChI

1S/C8H9N/c1-9-7-8-5-3-2-4-6-8/h2-7H,1H3/b9-7+

InChI key

HXTGGPKOEKKUQO-VQHVLOKHSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service