Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B102806

Sigma-Aldrich

Butyltriphenylphosphonium bromide

99%

Synonym(s):

NSC 59684, TBP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3P(Br)CH2CH2CH2CH3
CAS Number:
Molecular Weight:
399.30
Beilstein/REAXYS Number:
3580224
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

240-243 °C (lit.)

SMILES string

[Br-].CCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C22H24P.BrH/c1-2-3-19-23(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22;/h4-18H,2-3,19H2,1H3;1H/q+1;/p-1

InChI key

IKWKJIWDLVYZIY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Free radical 5-exo-dig cyclization
  • Intramolecular hydroacylalkoxylation
  • Wittig reactions for stereoselective synthesis of alkenes
  • Dimerization of α-olefins
  • Alkylidenation of hydrazides for synthesis of indoles
  • Tandem cyclization and 1,2-thiolate shift of nitrogen ylides

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Wang et al.
Journal of the American Society for Mass Spectrometry, 22(10), 1771-1783 (2011-09-29)
Fixed charge chemical modifications on peptides and proteins can impact fragmentation behaviors in tandem mass spectrometry (MS/MS). In this study, we employed a thiol-specific cationic alkylation reagent, (4-iodobutyl)triphenylphosphonium (IBTP), to selectively modify cysteine thiol groups in mitochondrial proteome samples. Tandem
J E McCarthy et al.
The Biochemical journal, 196(1), 311-321 (1981-04-15)
1. Aerobic respiration by cells of Paracoccus dentrificans drives the uptake of the lipophilic cation butyltriphenylphosphonium. Anaerobiosis or addition of an uncoupler of oxidative phosphorylation (carbonyl cyanide p-trifluoromethoxyphenylhydrazone) results in efflux of the cation. Changes in the concentration of butyltriphenylphosphonium
C W Keevil et al.
Analytical biochemistry, 139(1), 228-236 (1984-05-15)
Polyvinyl chloride membrane electrodes sensitive to tetraphenyl phosphonium (TPP+), butyltriphenyl phosphonium ( bTPP +), and methyltriphenyl phosphonium ( mTPP +) ions have been compared for the determination of the electrical potential difference (delta psi) of the oral bacteria, Streptococcus mutans
A P Singh et al.
Archives of biochemistry and biophysics, 245(2), 436-445 (1986-03-01)
Equilibration of the butyltriphenyl phosphonium (BTPP+) cation into cytochrome c oxidase reconstituted proteoliposomes was measured potentiometrically. The maximum membrane potential (delta psi) generated by oxidase activity was estimated to lie between -65 and -90 mV, vesicle interior negative, when internal
M Makrinich et al.
Physical chemistry chemical physics : PCCP, 22(37), 21022-21030 (2020-07-24)
Solid-state nuclear magnetic resonance has long been used to probe atomic distances between nearby nuclear spins by virtue of the dipolar interaction. New technological advances have recently enabled simultaneous tuning of the radio-frequency resonance circuits to nuclei with close Larmor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service