Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-LNPCKMP

Sigma-Aldrich

Coupling Kit For 200nm Magnetic Particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

usage

sufficient for 10 reactions

availability

not available in Japan

storage temp.

2-8°C

General description

The 200 nm Coupling Kit makes conducting lateral flow tests and biomolecule separation (including cell separation) easier and more flexible. The Kit contains AnteoBind activated 200 nm magnetic particles that give you increased antibody binding capacity and functionality, while the included blocking buffer decreases background noise.

Reduce reagent preparation time; remove traditional surface preparation steps such as EDC and replace these steps with the 200 nm pre-activated magnetic particles provided. This Kit reduces aggregation and gives you the freedom and ability to produce multifunctional particles for diverse applications, including dual labelling.

For lateral flow tests, magnetic particles are easier to handle than gold. Magnetic separation removes the need to perform centrifugation and filtration concentration. Magnetic particles can provide greater sensitivity than gold during lateral flow tests.

Binding Capacity and Polydisperity Index
Binding Capacity: > 50 μg IgG/mg
Polydispersity Index (PdI)*: < 0.3

* The Polydispersity Index (PdI) is dimensionless and determined using Dynamic Light Scattering (DLS). The PdI is scaled such that values smaller than 0.05 are rarely seen and values greater than 0.7 indicate that the sample has a very broad size distribution and poor monodispersity.

Application

Particle based Immunoassays, Lateral Flow, Bioseparations and Immunoprecipitation

Features and Benefits

  • 200 nm magnetic particles activated with water based AnteoBind
  • Easier and more flexible lateral flow tests
  • Easier and more flexible biomolecule separation
  • Increase antibody functionality
  • Reduce reagent preparation time by 3-4 hours
  • Highly monodispersed
  • Low background noise
  • Kit reduces aggregation
  • For diverse applications, including dual labelling
  • High reproducibility between experiments
  • Higher binding capacity than the Coupling Kit, 1 μm Magnetic Particles per mass of particles, potentially leading to a higher yield

Legal Information

6538 is a trademark of American Type Culture Collection
AnteoBind is a trademark of AnteoTech Ltd
Anteobind is a trademark of AnteoTech Ltd

Kit Components Only

Product No.
Description

  • Activated 200 nm Magnetic Particles

  • Coupling Buffer

  • Blocking Buffer

  • Storage Buffer

  • Low Binding Microcentrifuge Tubes

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service