ALD01830
N-(1-(benzyloxy)-2-methylpropan-2-yl)-4-fluoroaniline
About This Item
storage temp.
2-8°C
SMILES string
FC1=CC=C(NC(C)(C)COCC2=CC=CC=C2)C=C1
Application
related product
hcodes
Hazard Classifications
Aquatic Chronic 4
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service