ALD01728
4-((2-methypentan-2-yl)amino)phenyltrifluoromethanesulfonate
About This Item
storage temp.
2-8°C
SMILES string
CCCC(C)(C)NC1=CC=C(OS(=O)(C(F)(F)F)=O)C=C1
InChI
1S/C13H18F3NO3S/c1-4-9-12(2,3)17-10-5-7-11(8-6-10)20-21(18,19)13(14,15)16/h5-8,17H,4,9H2,1-3H3
InChI key
HZGQDLXAMQHXQC-UHFFFAOYSA-N
Application
related product
hcodes
Hazard Classifications
Aquatic Chronic 4
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service