Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00514

Sigma-Aldrich

Serine Hydrolase Inhibitor-3

Synonym(s):

tert-Butyl methyl(2-(1-methyl-3-(pyridin-2-yl)ureido)ethyl)carbamate, SHI-3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N4O3
CAS Number:
Molecular Weight:
308.38
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

form

liquid

SMILES string

O=C(N(CCN(C(OC(C)(C)C)=O)C)C)NC1=NC=CC=C1

InChI

1S/C15H24N4O3/c1-15(2,3)22-14(21)19(5)11-10-18(4)13(20)17-12-8-6-7-9-16-12/h6-9H,10-11H2,1-5H3,(H,16,17,20)

InChI key

AWNWUQFQKGBCMO-UHFFFAOYSA-N

Application

Serine hydrolase inhibitors are small molecule fragments that react in vitro with the serine hydrolase class of proteins. They are amenable to further modification to create novel inhibitors of this class of protein.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katerina Otrubova et al.
Bioorganic & medicinal chemistry letters, 24(16), 3807-3813 (2014-07-20)
Two libraries of modestly reactive ureas containing either electron-deficient acyl anilines or acyl pyrazoles were prepared and are reported as screening libraries for candidate serine hydrolase inhibitors. Within each library is a small but powerful subset of compounds that serve

Related Content

Boger Lab utilizes nature's solutions in complex natural products, extending them through rational design for improved biological outcomes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service