About This Item
description
Fp: >230°F
form
liquid
refractive index
n20/D 1.478
density
1.360 g/mL at 25 °C
SMILES string
O=S(OC1=C(OC)C=CC=C1)(F)=O
InChI
1S/C7H7FO4S/c1-11-6-4-2-3-5-7(6)12-13(8,9)10/h2-5H,1H3
Inchi Key
IHDUUYDISDOOKJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service