Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00410

Sigma-Aldrich

4-(methylsulfonyl)phenyl sulfofluoridate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7FO5S2
Molecular Weight:
254.26
UNSPSC Code:
12352100
PubChem Substance ID:

description

fp: >230°F

form

powder

mp

79-81 °C

SMILES string

O=S(OC1=CC=C(S(=O)(C)=O)C=C1)(F)=O

InChI

1S/C7H7FO5S2/c1-14(9,10)7-4-2-6(3-5-7)13-15(8,11)12/h2-5H,1H3

InChI key

OSOIRHOFNJYGET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The following aryl fluorosulfate can be utilized as a cross-coupling partner in Suzuki-Miyaura reactions. The standard reaction conditions for these palladium-catalyzed C-C bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service