Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00384

Sigma-Aldrich

Hydroquinone bisfluorosulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4F2O6S2
CAS Number:
Molecular Weight:
274.22
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

SMILES string

FS(OC1=CC=C(OS(F)(=O)=O)C=C1)(=O)=O

InChI

1S/C6H4F2O6S2/c7-15(9,10)13-5-1-2-6(4-3-5)14-16(8,11)12/h1-4H

InChI key

OEJXDBMJMKHHMQ-UHFFFAOYSA-N

Application

The following aryl fluorosulfate can be utilized as a cross-coupling partner in Suzuki-Miyaura reactions. The standard reaction conditions for these palladium-catalyzed C-C bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service