Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00376

Sigma-Aldrich

Shi-Yu MPAA Ligand

Synonym(s):

N-[[[(1R,2S,5R)-5-methyl-2-(1-methylethyl)cyclohexyl]oxy]carbonyl]-L-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H31NO4
CAS Number:
Molecular Weight:
313.43
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: C-H Activation
reagent type: catalyst
reagent type: ligand
reaction type: Peptide Synthesis

functional group

amine
carboxylic acid

storage temp.

2-8°C

SMILES string

O=C(O)[C@@H](NC(O[C@H]1[C@H](C(C)C)CC[C@@H](C)C1)=O)CC(C)C

InChI

1S/C17H31NO4/c1-10(2)8-14(16(19)20)18-17(21)22-15-9-12(5)6-7-13(15)11(3)4/h10-15H,6-9H2,1-5H3,(H,18,21)(H,19,20)/t12-,13+,14+,15-/m1/s1

InChI key

JDNKQMWGCKCHBF-CBBWQLFWSA-N

Application

Shi-Yu MPAA Ligand is a monoprotected α-amino acid that can be used as an effective chiral ligand in the asymmetric palladium-catalyzed alkylation reactions. It is also utilized in the alkylation of diphenyl(2-pyridyl)methane and isopropyl pyridines with alkyl boronic acids via enantioselective activation of C(sp2)-H and C(sp3)-H bonds, respectively.[1]
The Shi-Yu MPAA Ligand has been reported by Yu and coworkers to promote the enantioselective C-H activation of cyclopropanes. This transformation is carried out through the coupling of organoboron reagents using palladium-mediated catalysis.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd(II)-catalyzed enantioselective activation of C(sp2)-H and C(sp3)-H bonds using monoprotected amino acids as chiral ligands.
Bing-Feng Shi et al.
Angewandte Chemie (International ed. in English), 47(26), 4882-4886 (2008-05-20)

Related Content

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service