Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ALD00234

Sigma-Aldrich

Sodium tetrahydrofuransulfinate

≥90% (H-NMR)

Synonym(s):

Baran TFHS-Na Reagent, Sodium oxolane-3-sulfinate, Baran THFS-Na Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NaO3S
CAS Number:
Molecular Weight:
158.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥90% (H-NMR)

form

solid

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

SMILES string

O=S([O-])C1CCOC1.[Na+]

InChI

1S/C4H8O3S.Na/c5-8(6)4-1-2-7-3-4;/h4H,1-3H2,(H,5,6);/q;+1/p-1

InChI key

IHAPRSZBTHYXPY-UHFFFAOYSA-M

Application

The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service