Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00222

Sigma-Aldrich

4-Bromo-2-chlorobenzenesulfonyl fluoride

95%

Synonym(s):

4-Bromo-2-chlorobenzenesulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3BrClFO2S
CAS Number:
Molecular Weight:
273.51
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

reaction suitability

reaction type: click chemistry

mp

58-63 °C

SMILES string

FS(C1=C(Cl)C=C(Br)C=C1)(=O)=O

InChI

1S/C6H3BrClFO2S/c7-4-1-2-6(5(8)3-4)12(9,10)11/h1-3H

InChI key

VHKMFTSOYSGAJW-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service