ALD00206
3,5-Dimethylbenzenesulfonyl fluoride
95% (H-NMR)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H9FO2S
CAS Number:
Molecular Weight:
188.22
UNSPSC Code:
12352100
PubChem Substance ID:
assay
95% (H-NMR)
form
solid
reaction suitability
reaction type: click chemistry
mp
35-40 °C
SMILES string
CC1=CC(C)=CC(S(F)(=O)=O)=C1
InChI
1S/C8H9FO2S/c1-6-3-7(2)5-8(4-6)12(9,10)11/h3-5H,1-2H3
InChI key
USDUCJJANRAWTQ-UHFFFAOYSA-N
Application
Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service