ALD00154
4,5-Dichloro-2-thiophenesulfonyl fluoride
95%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4HCl2FO2S2
CAS Number:
Molecular Weight:
235.08
UNSPSC Code:
12352100
PubChem Substance ID:
assay
95%
form
solid
reaction suitability
reaction type: click chemistry
mp
27-32 °C
SMILES string
O=S(C1=CC(Cl)=C(Cl)S1)(F)=O
InChI
1S/C4HCl2FO2S2/c5-2-1-3(10-4(2)6)11(7,8)9/h1H
InChI key
CTFSVTLSIBOOMZ-UHFFFAOYSA-N
Application
Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service