Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ALD00072

Sigma-Aldrich

2-Bromo-4-fluorobenzenesulfonyl fluoride

95% (GC)

Synonym(s):

2-Bromo-4-fluorobenzenesulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3BrF2O2S
Molecular Weight:
257.05
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95% (GC)

form

solid

reaction suitability

reaction type: click chemistry

mp

32-36 °C

SMILES string

O=S(C1=CC=C(F)C=C1Br)(F)=O

InChI

1S/C6H3BrF2O2S/c7-5-3-4(8)1-2-6(5)12(9,10)11/h1-3H

InChI key

ONERXDMQVCUGBT-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service