Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00064

Sigma-Aldrich

2,5-Dimethoxybenzenesulfonyl fluoride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9FO4S
CAS Number:
Molecular Weight:
220.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

95%

form

solid

reaction suitability

reaction type: click chemistry

mp

108-113 °C

SMILES string

FS(C1=C(OC)C=CC(OC)=C1)(=O)=O

InChI

1S/C8H9FO4S/c1-12-6-3-4-7(13-2)8(5-6)14(9,10)11/h3-5H,1-2H3

InChI key

HOHJQDUMZBLDDY-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In collaboration with K. Barry Sharpless and coworkers, we offer a variety of sulfonyl fluoride reagents that undergo a “Click II” reaction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service