Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00020

Sigma-Aldrich

2-((1-(2-Oxo-2-phenylethyl)-1H-pyrrol-2-yl)methylene)malononitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N3O
Molecular Weight:
261.28
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

storage temp.

2-8°C

SMILES string

O=C(CN1C(C=C(C#N)C#N)=CC=C1)C2=CC=CC=C2

InChI

1S/C16H11N3O/c17-10-13(11-18)9-15-7-4-8-19(15)12-16(20)14-5-2-1-3-6-14/h1-9H,12H2

InChI key

WRVKWUUMYYUBNT-UHFFFAOYSA-N

Application

Building block is employed in the enantioselective synthesis of 5, 6-dihydroindolizines through a cascade sequence initiated by (S)-(–)-α, α-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether (Aldrich 677183).

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service