Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00014

Sigma-Aldrich

Ethyl (E)-2-(5-methyl-2-oxobenzofuran-3(2H)-ylidene)acetate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12O4
CAS Number:
Molecular Weight:
232.23
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

flakes

storage temp.

2-8°C

SMILES string

CC1=CC2=C(OC(/C2=C/C(OCC)=O)=O)C=C1

InChI

1S/C13H12O4/c1-3-16-12(14)7-10-9-6-8(2)4-5-11(9)17-13(10)15/h4-7H,3H2,1-2H3/b10-7+

InChI key

YBQXFHKKDZKMKA-JXMROGBWSA-N

Application

Building block used as a substrate in phosphine-catalyzed domino sequences. This versatile compound can be used to synthesize a wide scope of spirocyclic benzofuranone scaffolds.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service