Key Documents
ACE1353010
Ace stir motor controller
AC/DC input 120 V AC, 60 Hz, 10 A
About This Item
UNSPSC Code:
41106514
AC/DC input
120 V, US 3-pin plug
packaging
pack of 1 ea
manufacturer/tradename
Ace Glass 13530-10
General description
Rated 1 hp for use with most stirrer motors. Not for use with shunt wound DC motors. A compact, lightweight, solid state, ring stand mountable control unit, both AC and DC outlets. Pulsed AC gives better low speed torque than voltage dropping devices such as rheostat or transformer.
AC output: 120 volts, 10 amps. DC motors give better all- around performance than most other solid state controllers because rectification is balanced full-wave instead of half-wave power. DC output: 0-120 volts, 6 amps. Features top Fwd-Off-Rev. motor switch.
Use motor with Ace stir motor ACE1364909, sold separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)