Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A63007

Sigma-Aldrich

4-Amino-3-methylbenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
151.16
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

169-171 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cc1cc(ccc1N)C(O)=O

InChI

1S/C8H9NO2/c1-5-4-6(8(10)11)2-3-7(5)9/h2-4H,9H2,1H3,(H,10,11)

InChI key

NHFKECPTBZZFBC-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weixiang Liu et al.
Carbohydrate polymers, 160, 97-105 (2017-01-25)
A novel type of O-carboxymethyl chitosan Schiff bases (O-CSPX) was synthesized via a condensation reaction. After the coordination reaction of cupric ions, Cu(II) complexes (O-CSPX-Cu) were achieved. The theoretical structure of O-CSPX-Cu calculated by Gaussian 09 reveals that the copper
Chia-Ju Tsai et al.
Journal of chromatography. A, 1524, 29-36 (2017-10-11)
Detection of polar organic compounds (POCs) using gas chromatography (GC) is not straightforward due to high polarity, hydrophilicity, and low volatility of POCs. In this study, we report a tandem microwave-assisted derivatization method combined with salting-out assisted liquid-liquid microextraction (SALLME)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service