Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3384

Sigma-Aldrich

5(4)-Aminoimidazole-4(5)-carboxamidoxime dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N5O · 2HCl
CAS Number:
Molecular Weight:
214.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

storage temp.

−20°C

SMILES string

Cl.Nc1nc[nH]c1C(=N)NO

InChI

1S/C4H7N5O.2ClH/c5-3-2(4(6)9-10)7-1-8-3;;/h1,10H,5H2,(H2,6,9)(H,7,8);2*1H

InChI key

VUUHBIHWGRRJKD-UHFFFAOYSA-N

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service