Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A22807

Sigma-Aldrich

1-Acetyl-3-thiosemicarbazide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7N3OS
CAS Number:
Molecular Weight:
133.17
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

powder

mp

165-168 °C (lit.)

SMILES string

CC(=O)NNC(N)=S

InChI

1S/C3H7N3OS/c1-2(7)5-6-3(4)8/h1H3,(H,5,7)(H3,4,6,8)

InChI key

NSIMQTOXNOFWBP-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aeshah Hassan Alamri
Materials (Basel, Switzerland), 13(21) (2020-11-12)
Experimental insights into the synergistic effect of 1-acetyl-3-thiosemicarbazide (AST) and iodide ions on the corrosion of C1018 carbon steel in 1 M HCl solution were investigated using open-circuit potential (OCP), linear polarization resistance (LPR), electrochemical frequency modulation (EFM), potentiodynamic polarization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service