Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A104

Sigma-Aldrich

Acenaphthene

97%

Synonym(s):

1,8-Ethylenenaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10
CAS Number:
Molecular Weight:
154.21
Beilstein/REAXYS Number:
386081
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.32 (vs air)

vapor pressure

10 mmHg ( 131 °C)

assay

97%

form

solid

bp

279 °C (lit.)

mp

90-94 °C (lit.)

SMILES string

C1Cc2cccc3cccc1c23

InChI

1S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6H,7-8H2

InChI key

CWRYPZZKDGJXCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

257.0 °F - closed cup

flash_point_c

125.0 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanzheng Gao et al.
Journal of environmental quality, 40(2), 653-656 (2011-04-28)
The subcellular process and distribution of polycyclic aromatic hydrocarbons (PAHs) in arbuscular mycorrhizal plants remains to be elucidated. This work used a greenhouse experiment to show that, accompanied by the apoplastic and symplastic water movement through the root, acenaphthene (ACE)
Ningjing Hu et al.
Environmental science and pollution research international, 18(2), 163-172 (2010-06-26)
The levels and possible sources of 16 polycyclic aromatic hydrocarbons (PAHs) were investigated in the surface sediments of Liaodong Bay, Bohai Sea, China. The sum of 16 PAHs (∑PAH(16)) concentrations varied from 144.5 to 291.7 ng/g, with a mean value
Di Wu et al.
Biomedical and environmental sciences : BES, 20(4), 291-294 (2007-10-24)
To investigate the oxidation of acenaphthene (Ace), a polycyclic aromatic hydrocarbon (PAH) with a saturated C-C bond by ozone and to characterize the intermediate products of ozonation. Ozone was generated from filtered dry oxygen by an ozone generator and continually
Rachel R Butorac et al.
Molecules (Basel, Switzerland), 16(4), 3168-3178 (2011-04-19)
The syntheses of four new bis(imino)acenaphthene (BIAN) imidazolium chlorides are reported, three of which have been structurally characterized. The synthesis of a new, structurally authenticated BIAN ligand is also described. We report the results of the use of these BIAN
Li Lu et al.
Environmental science and pollution research international, 19(5), 1515-1523 (2011-10-15)
Cationic surfactants are common in soils because of their use in daily cosmetic and cleaning products, and their use as a soil amendment for the mitigation and remediation of organic contaminated soils has been proposed. Such surfactant may affect the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service