Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

97240

Sigma-Aldrich

Z-Trp-OH

97%

Synonym(s):

N-Cbz-L-tryptophan, Nα-Carbobenzyloxy-L-tryptophan, Z-L-tryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O4
CAS Number:
Molecular Weight:
338.36
Beilstein/REAXYS Number:
96743
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

97%

optical activity

[α]20/D +2.9±0.3°, c = 3% in acetic acid

reaction suitability

reaction type: solution phase peptide synthesis

mp

124-127 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)OCc3ccccc3

InChI

1S/C19H18N2O4/c22-18(23)17(10-14-11-20-16-9-5-4-8-15(14)16)21-19(24)25-12-13-6-2-1-3-7-13/h1-9,11,17,20H,10,12H2,(H,21,24)(H,22,23)/t17-/m0/s1

InChI key

AHYFYYVVAXRMKB-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S R Vigna et al.
Brain research, 343(2), 394-397 (1985-09-23)
The abilities of the pancreatic cholecystokinin (CCK) receptor antagonists dibutyryl cyclic GMP, proglumide, benzotript, CBZ-tryptophan, CBZ-cysteine and CCK-27-32-amide to inhibit CCK binding to its receptor in the pancreas and brain of mice and guinea pigs was examined. In both species
R Magous et al.
Biochimica et biophysica acta, 845(2), 158-162 (1985-05-30)
Benzotript (N-p-chlorobenzoyl-L-tryptophan) has been shown to be a receptor-antagonist in vivo and in vitro for peptides from the gastrin family. In the present study, we examine tryptophan, and some of its N- and C-acylated derivatives, as well as some phenylalanine
Jason Lejeune et al.
Biosensors & bioelectronics, 25(3), 604-608 (2009-03-27)
Changes detected in the imprinting effect by OMNiMIPs imprinted with multiple templates appear to be a function of the maximum template loading. Below the maximum template loading, the polymers imprinted with multiple compounds provide molecular recognition close to the polymers
Mariangela Ceruso et al.
Biochimica et biophysica acta, 1824(2), 303-310 (2011-11-01)
The chemical shift of the carboxylate carbon of Z-tryptophan is increased from 179.85 to 182.82 ppm and 182.87 ppm on binding to thermolysin and stromelysin-1 respectively. The chemical shift of Z-phenylalanine is also increased from 179.5 ppm to 182.9 ppm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service