96304
N-Z-L-Homoserine lactone
≥98.0% (HPLC)
Synonym(s):
(S)-α-(Z-Amino)-γ-butyrolactone
About This Item
assay
≥98.0% (HPLC)
optical activity
[α]20/D −32±2°, c = 1.7% in methanol
reaction suitability
reaction type: solution phase peptide synthesis
mp
127-132 °C
application(s)
peptide synthesis
SMILES string
O=C(N[C@H]1CCOC1=O)OCc2ccccc2
InChI
1S/C12H13NO4/c14-11-10(6-7-16-11)13-12(15)17-8-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H,13,15)/t10-/m0/s1
Inchi Key
FKWDZIFOVOUDAG-JTQLQIEISA-N
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service