Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95921

Sigma-Aldrich

Eumulgin® B25

~100% active ingredients basis

Synonym(s):

Macrogol (25)-cetostearyl ether, Polyethylene glycol 1100 mono(hexadecyl/octadecyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103

description

non-ionic

assay

~100% active ingredients basis

technique(s)

protein quantification: suitable

impurities

≤1 mg/kg ethylene oxide
≤10 mg/kg dioxane

solubility

H2O: 1 g/10 mL, clear, colorless

Looking for similar products? Visit Product Comparison Guide

Application

Eumulgin® B 25 is an oil-in-water emulsifier that is used for the production of ointments, creams, liquid emulsions, gels and other cosmetic formulations. It is supplied in the form of free-flowing, non-dusting micro-granules, and is an efficient protective colloid that can increase the stability of dispersed systems. This research grade product is intended for use in R&D only.

Legal Information

Eumulgin is a registered trademark of Cognis IP Management GmbH

related product

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

469.4 °F - closed cup

flash_point_c

243 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia D Stan et al.
Cancer research, 68(18), 7661-7669 (2008-09-17)
Withaferin A (WA) is derived from the medicinal plant Withania somnifera, which has been safely used for centuries in Indian Ayurvedic medicine for treatment of different ailments. We now show, for the first time, that WA exhibits significant activity against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service