Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95528

Sigma-Aldrich

Dipropyl sulfate

allkylating agent, ≥97.0% (GC)

Synonym(s):

Propyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2O)2SO2
CAS Number:
Molecular Weight:
182.24
Beilstein/REAXYS Number:
1705816
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.413

density

1.109 g/mL at 20 °C (lit.)

SMILES string

CCCOS(=O)(=O)OCCC

InChI

1S/C6H14O4S/c1-3-5-9-11(7,8)10-6-4-2/h3-6H2,1-2H3

InChI key

JYCKNDWZDXGNBW-UHFFFAOYSA-N

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service