Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95077

Sigma-Aldrich

1-Vinyl-1,2,4-triazole

≥97.0%

Synonym(s):

1-Ethenyl-1H-1,2,4-triazole, 1-Vinyl-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3
CAS Number:
Molecular Weight:
95.10
Beilstein/REAXYS Number:
605827
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0%

form

liquid

refractive index

n20/D 1.511

density

1.099 g/mL at 20 °C (lit.)

SMILES string

C=Cn1cncn1

InChI

1S/C4H5N3/c1-2-7-4-5-3-6-7/h2-4H,1H2

InChI key

ITUNZCVRYICLQM-UHFFFAOYSA-N

Application

1-Vinyl-1,2,4-triazole is mainly used in the preparation of poly(1-vinyl-1,2,4-triazole) and its copolymers.[1][2][3]

Introduction of 1-vinyl-1,2,4-triazole in polysaccharide based semi-degradable hydrogel to increase the equilibrium water content and endow hydrogels with anti-bacterial and anti-inflammatory abilities has also been reported.[4]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of poly (1-vinyltriazole)-grafted superparamagnetic iron oxide nanoparticles.
Deligoz H, et al.
Synt. Metals, 162(7-8), 590-597 (2012)
A high strength semi-degradable polysaccharide-based hybrid hydrogel for promoting cell adhesion and proliferation.
Wang H, et al.
J. Mater. Sci., 53(9), 6302-6312 (2018)
Phosphoric acid-doped poly (1-vinyl-1, 2, 4-triazole) as water-free proton conducting polymer electrolytes.
Celik S U, et al.
Solid State Ionics, 179(19-20), 683-688 (2008)
Proton?conducting blend membranes of crosslinked poly (vinyl alcohol)?sulfosuccinic acid ester and poly (1?vinyl?1, 2, 4?triazole) for high temperature fuel cells.
Boroglu M S, et al.
Polym. Eng. Sci., 53(1), 153-158 (2013)
Jian-Ke Sun et al.
Nature communications, 9(1), 1717-1717 (2018-05-02)
Soft actuators with integration of ultrasensitivity and capability of simultaneous interaction with multiple stimuli through an entire event ask for a high level of structure complexity, adaptability, and/or multi-responsiveness, which is a great challenge. Here, we develop a porous polycarbene-bearing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service