Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94905

Sigma-Aldrich

2-[Bis(2,4-di-tert-butyl-phenoxy)phosphinooxy]-3,5-di(tert-butyl)phenyl-palladium(II) chloride dimer

~12% Pd basis

Synonym(s):

Bedford Catalyst, Bis[2-(bis(2,4-di-tert-butylphenoxy)phosphino-κP-oxy)-3,5-di-tert-butylphenyl-κC]di-μ-chloro-dipalladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C84H124Cl2O6P2Pd2
CAS Number:
Molecular Weight:
1575.57
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

crystals

reaction suitability

reagent type: catalyst
core: palladium

concentration

~12% Pd

SMILES string

CC(C)(C)c1ccc(OP(Oc2ccc(cc2C(C)(C)C)C(C)(C)C)Oc3c(cc(cc3C(C)(C)C)C(C)(C)C)[Pd-]4[Cl+][Pd-]([Cl+]4)c5cc(cc(c5OP(Oc6ccc(cc6C(C)(C)C)C(C)(C)C)Oc7ccc(cc7C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C

InChI

1S/2C42H62O3P.2Cl.2Pd/c2*1-37(2,3)28-19-22-34(31(25-28)40(10,11)12)43-46(44-35-23-20-29(38(4,5)6)26-32(35)41(13,14)15)45-36-24-21-30(39(7,8)9)27-33(36)42(16,17)18;;;;/h2*19-23,25-27H,1-18H3;;;;/q;;2*+1;2*-1

InChI key

HPRPCQXYHQYGHY-UHFFFAOYSA-N

Application

Catalyst for:
  • Racemic and asymmetric sequential aldol condensations / addition reactions
  • Miyaura borylations
  • Lewis acid catalysis
  • Allylation of aldehydes
  • Addition reactions
  • Alkoxy- and amidocarbonylations

Other Notes

Very active catalyst for coupling reactions proceeding in high turnover; it shows good air and thermal stability

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical Communications (Cambridge, England), 2095-2095 (1998)
Tetrahedron Letters, 39, 9793-9793 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service