Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94489

Sigma-Aldrich

1-Methyl-5-nitroimidazole

>97.0% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H5N3O2
CAS Number:
Molecular Weight:
127.10
Beilstein/REAXYS Number:
123550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

>97.0% (HPLC)

form

solid

mp

54-57 °C

functional group

nitro

SMILES string

Cn1cncc1[N+]([O-])=O

InChI

1S/C4H5N3O2/c1-6-3-5-2-4(6)7(8)9/h2-3H,1H3

InChI key

JLZXSFPSJJMRIX-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mónica Mendes et al.
International journal of molecular sciences, 20(24) (2019-12-11)
Electron transfer induced decomposition mechanism of nitroimidazole and a selection of analogue molecules in collisions with neutral potassium (K) atoms from 10 to 1000 eV have been thoroughly investigated. In this laboratory collision regime, the formation of negative ions was
S Pandeti et al.
The Journal of chemical physics, 150(1), 014302-014302 (2019-01-10)
Nitroimidazolic radiosensitizers are used in radiation therapy to selectively sensitize cancer cells deprived of oxygen, and the actual mechanism of radiosensitization is still not understood. Selecting five radiosensitizers (1-methyl-5-nitroimidazole, ronidazole, ornidazole, metronidazole, and nimorazole) with a common 5-nitroimidazolic ring with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service