Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92455

Sigma-Aldrich

S,S′-Trimethylene di(p-toluenethiosulfonate)

≥97.0% (HPLC)

Synonym(s):

1,3-Di(p-tosylthio)propane, 1,3-Propanediyl di(thiotosylate), Trimethylene di(thiotosylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2SCH2CH2CH2SSO2C6H4CH3
CAS Number:
Molecular Weight:
416.60
Beilstein/REAXYS Number:
2020157
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (HPLC)

form

crystals

mp

64-67 °C

functional group

disulfide

SMILES string

Cc1ccc(cc1)S(=O)(=O)SCCCSS(=O)(=O)c2ccc(C)cc2

InChI

1S/C17H20O4S4/c1-14-4-8-16(9-5-14)24(18,19)22-12-3-13-23-25(20,21)17-10-6-15(2)7-11-17/h4-11H,3,12-13H2,1-2H3

InChI key

QOICHLMIPNOKLN-UHFFFAOYSA-N

Other Notes

Reagent for the formation of dithianes (protected carbonyls) from activated methylene compounds[1][2][3][4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Suzuki et al.
Chemical & Pharmaceutical Bulletin, 34, 3135-3135 (1986)
A.K. Saksena et al.
Tetrahedron Letters, 22, 5227-5227 (1981)
R.B. Woodward et al.
Organic Syntheses, 54, 39-39 (1974)
J.E. McMurry et al.
The Journal of Organic Chemistry, 49, 3803-3803 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service