Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92040

Sigma-Aldrich

Triisopentylamine

technical, ≥90% (GC)

Synonym(s):

Triisoamylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2CH2]3N
CAS Number:
Molecular Weight:
227.43
Beilstein/REAXYS Number:
1720767
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.433

bp

265-270 °C (lit.)

density

0.782 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

CC(C)CCN(CCC(C)C)CCC(C)C

InChI

1S/C15H33N/c1-13(2)7-10-16(11-8-14(3)4)12-9-15(5)6/h13-15H,7-12H2,1-6H3

InChI key

QKVUSSUOYHTOFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

170.6 °F - closed cup

flash_point_c

77 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service