Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91934

Sigma-Aldrich

4-(Pentafluorosulfanyl)benzene-1,2-diamine

≥95% (HPLC)

Synonym(s):

(3,4-Diaminophenyl)sulfur pentafluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7F5N2S
CAS Number:
Molecular Weight:
234.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥95% (HPLC)

form

solid

suitability

conforms to structure for Proton NMR spectrum

storage temp.

2-8°C

SMILES string

NC1=C(N)C=C(S(F)(F)(F)(F)F)C=C1

InChI

1S/C6H7F5N2S/c7-14(8,9,10,11)4-1-2-5(12)6(13)3-4/h1-3H,12-13H2

InChI key

UIJLRAJFHPRCAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service