Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91682

Sigma-Aldrich

N-(3-Hydroxypropyl)trifluoroacetamide

≥98.0% (GC)

Synonym(s):

3-(Trifluoroacetamido)-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8F3NO2
CAS Number:
Molecular Weight:
171.12
Beilstein/REAXYS Number:
4969338
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (GC)

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.406

bp

120-121 °C/0.03 mmHg (lit.)

density

1.355 g/mL at 20 °C (lit.)

SMILES string

OCCCNC(=O)C(F)(F)F

InChI

1S/C5H8F3NO2/c6-5(7,8)4(11)9-2-1-3-10/h10H,1-3H2,(H,9,11)

InChI key

LOTWHTFZRDJSCL-UHFFFAOYSA-N

Other Notes

N-Protected 3-aminopropanol

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zoltán Dienes et al.
The Journal of organic chemistry, 61(20), 6958-6970 (1996-10-04)
The BF(3).Et(2)O-promoted Diels-Alder addition of 1-acetylvinyl RADO(Et)-ate (RADO(Et)-ate = 3-ethyl-2-oxo-6,8-dioxa-3-azabicyclo[3.2.1]octane-7-exo-carboxylate) to 1-(dimethoxymethyl)-2,3,5,6-tetramethylidene-7-oxabicyclo[2.2.1]heptane led to one major monoadduct that added to 1,2-didehydrobenzene and was converted into (-)-4-demethoxy-7-deoxydaunomycinone and (2R)-12-acetoxy-2-acetyl-5-(bromomethyl)-1,2,3,4-tetrahydronaphthacen-2-yl RADO(Et)-ate. The latter compound was used to construct (8R)-8-acetyl-6,8-dihydroxy-11-[[(3'-[(aminopropyl)oxy]-, -4'-[(aminobutyl)oxy], and -5'-[(aminopentyl)oxy]methyl]-7,8,9,10-tetrahydronaphthacene-5,12-dione

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service