Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91671

Sigma-Aldrich

2-(Trifluoroacetoxy)pyridine

≥98.0%

Synonym(s):

2-Pyridyl trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4F3NO2
CAS Number:
Molecular Weight:
191.11
Beilstein/REAXYS Number:
3607316
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0%

form

liquid

refractive index

n20/D 1.421 (lit.)
n20/D 1.422

bp

60 °C/0.3 mmHg (lit.)

density

1.356 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)Oc1ccccn1

InChI

1S/C7H4F3NO2/c8-7(9,10)6(12)13-5-3-1-2-4-11-5/h1-4H

InChI key

BJCXMOBNIFBHBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Mild trifluoroacetylating agent for amines, alcohols and phenols, no base needs to be added[1]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

122.0 °F - closed cup

flash_point_c

50 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Keumi et al.
Bulletin of the Chemical Society of Japan, 63, 2252-2252 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service