915769
Nafion™ perfluorinated membrane
Nafion™ 117, thickness × L × W 0.007 in. × 0.61 m × 2.5 m, sheet
About This Item
Recommended Products
form
sheet
eq. wt.
1,100
manufacturer/tradename
Nafion™ 117
thickness × L × W
0.007 in. × 0.61 m × 2.5 m
solubility
H2O: insoluble
H2O: insoluble
InChI
1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);
InChI key
FOYUGSIADQEOEK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Nafion™ is a perfluorinated ion-exchange membrane which has a wide variety of commercial uses. The acidic (hydrogen ion) form and derivatives are solid, superacid catalysts useful in a wide variety of synthetic applications.
Packaging
· Keep pieces sealed and protected from general exposure will reduce the effect. Recommend long term storage in air-tight bags.
· Discoloration does not affect performance and can be removed by soaking in dilute nitric acid.
- Keep pieces sealed and protected from general exposure will reduce the effect. Recommend long term storage in air-tight bags.
- Discoloration does not affect performance and can be removed by soaking in dilute nitric acid.
Legal Information
related product
Storage Class
11 - Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service