Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91550

Sigma-Aldrich

Tridecanoyl chloride

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11COCl
CAS Number:
Molecular Weight:
232.79
Beilstein/REAXYS Number:
1767828
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥99.0% (GC)

refractive index

n20/D 1.447

density

0.914 g/mL at 20 °C

SMILES string

CCCCCCCCCCCCC(Cl)=O

InChI

1S/C13H25ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3

InChI key

FJRPWCNFWGBGOF-UHFFFAOYSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service