Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

915300

Sigma-Aldrich

Platinum nanospheres

citrate functionalized, 70±6 nm diameter, 0.05 mg/mL (4 mM aqueous sodium citrate buffer)

Synonym(s):

Platinum nanoparticles, PtNP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
UNSPSC Code:
12161600

form

nanospheres

concentration

0.05 mg/mL (4 mM aqueous sodium citrate buffer)

diameter

70±6 nm

functional group

citrate

storage temp.

2-8°C

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Platinum nanoparticles have many applications ranging from the automotive, chemical, and biomedical industries.[1][2][3]

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah Pedone et al.
Chemical Society reviews, 46(16), 4951-4975 (2017-07-12)
Oxidative stress-dependent inflammatory diseases represent a major concern for the population's health worldwide. Biocompatible nanomaterials with enzymatic properties could play a crucial role in the treatment of such pathologies. In this respect, platinum nanoparticles (PtNPs) are promising candidates, showing remarkable
Recent Advances in FePt Nanoparticles for Biomedicine.
Shi Y, et al.
Journal of Nanomaterials, 2015, 1-13 (2015)
Pt-based composite nanoparticles for magnetic, catalytic, and biomedical applications.
Liu Y, et al.
Journal of Materials Chemistry, 21, 12579-12587 (2011)

Articles

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service