Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91381

Sigma-Aldrich

3-Ethyl-1-phenyltriazene polymer-bound

purum, ≥95.0% (T), extent of labeling: ~1.0 mmol/g loading

Synonym(s):

4-(Ethylazoamino)phenoxymethyl-polystyrene, 4-(Ethyltriazeno)phenoxymethyl-polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

grade

purum

assay

≥95.0% (T)

form

powder

extent of labeling

~1.0 mmol/g loading

impurities

≤1.0% water

matrix

crosslinked with 2% DVB

particle size

100-200 mesh

storage temp.

2-8°C

Other Notes

Polymer bound reagent for the conversion of acids into ethyl esters[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernhard Erb et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 9(11), 2582-2588 (2003-06-10)
Starting from polystyrene, a simple four-step synthesis of polymer-supported alkyltriazenes (alkyl=Me, Et, benzyl) is described. With this synthesis, a loading capacity of 2.2 mmol g(-1) can be reached. The most prominent application of these polymer-supported reagents is the rapid, highly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service