About This Item
assay
≥97% (GC)
refractive index
n20/D 1.470
bp
224-226 °C (lit.)
density
1.389 g/mL at 20 °C (lit.)
SMILES string
CCOC(=O)CC(=O)C(Cl)(Cl)Cl
InChI
1S/C6H7Cl3O3/c1-2-12-5(11)3-4(10)6(7,8)9/h2-3H2,1H3
InChI key
XGIRWPYENXRJMZ-UHFFFAOYSA-N
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service