Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

908398

Sigma-Aldrich

[(S)-(-)-5,5′-Bis(ditolylphosphino)-4,4′-binaphthalene][4-cyano-3-nitrobenzenecarboxylato][1,2,3-η-2-propenyl]iridium(III)

Synonym(s):

Krische-Zbieg Catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H47IrN2O4P2
CAS Number:
Molecular Weight:
1102.18
UNSPSC Code:
12352200

form

powder

mp

230 °C (decomp)

storage temp.

2-8°C

Application

[(S)-(-)-5,5′-Bis(ditolylphosphino)-4,4′-binaphthalene][4-cyano-3-nitrobenzenecarboxylato][1,2,3-η-2-propenyl]iridium(III) is a catalyst developed in the Krische lab for the regio- and enantioselective amination of alkyl-substituted allylic acetates with aryl amines.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung Wook Kim et al.
Journal of the American Chemical Society, 141(1), 671-676 (2018-12-21)
The air- and water-stable π-allyliridium C,O-benzoate modified by ( S)-tol-BINAP, ( S)-Ir-II, catalyzes highly regio- and enantioselective Tsuji-Trost-type aminations of racemic branched alkyl-substituted allylic acetates using primary or secondary (hetero)aromatic amines. Specifically, in the presence of ( S)-Ir-II (5 mol%)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service