Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90808

Sigma-Aldrich

Tributylmethylphosphonium chloride polymer-bound

extent of labeling: ~1.4 mmol/g Cl- loading

Synonym(s):

Methyltributylphosphonium chloride polymer-bound, Tributylphosphoniomethyl-polystyrene crosslinked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:

extent of labeling

~1.4 mmol/g Cl- loading

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

Other Notes

Immobilized tetraalkylphosphonium salt with high catalytic activity in PT reactions [1][2][3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Tomoi et al.
Journal of the American Chemical Society, 103, 3821-3821 (1981)
F. Montanari et al.
The Journal of Organic Chemistry, 48, 199-199 (1983)
S.L. Regen et al.
The Journal of Organic Chemistry, 46, 2511-2511 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service