Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

906999

Sigma-Aldrich

Graphene oxide

anode grade, 10 mg/mL

Synonym(s):

1 wt% graphene oxide in water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CxOyHz
UNSPSC Code:
12352119

description

Grade: anode additive grade

form

dispersion

concentration

10 mg/mL

storage temp.

2-8°C

application

This anode grade graphene oxide is specifically optimized as starting material for additives in lithium ion battery anodes. Graphene oxide and reduced graphene oxide have been heavily used as additives to wrap active lithium ion battery materials.[1][2][3] The mechanical property of graphene based sheets can provide structure integrity for the active material during charge/discharge induced huge volume change[2][3][4], and thus results in improved cycle life. Furthermore, graphene and reduced graphene oxide can give extra benifits such as fast charge rate and large capacity, due to their high conductivity and electrochemical surface area.[2]

Caution

Shake or sonicate before use.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Si nanoparticles/graphene composite membrane for high performance silicon anode in lithium ion batteries.
Zhongping L, et al.
Carbon, 98, 373-380 (2016)
Cobalt phosphide nanocages encapsulated with graphene as ultralong cycle life anodes for reversible lithium storage.
Mengmeng D, et al.
Research on Chemical Intermediates, 44(12), 7847-7859 (2018)
Fe2O3 particles enwrapped by graphene with excellent cyclability and rate capability as anode materials for lithium ion batteries.
Xiao W, et al.
Applied Surface Science, 266, 148-154 (2013)
Dan Thien Nguyen et al.
ACS applied materials & interfaces, 5(21), 11234-11239 (2013-10-12)
We present the first report on carbon fiber-interwoven amorphous nano-SiOx/graphene prepared by a simple and facile room temperature synthesis of amorphous SiOx nanoparticles using silica, followed by their homogeneous dispersion with graphene nanosheets and carbon fibers in room temperature aqueous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service