The extracellular matrix (ECM), comprised of proteins and other growth factors, plays a pivotal role in directing and mediating cellular activities in a tissue or an organ. Decellularized extracellular matrix (dECM) could not only replicate natural ECM microenvironment, but also provide tissue specific compositions and topology. dECM bioink is capable of providing optimized tissue-specific microenvironment conducive to the growth of 3D structured tissues. Decellularized ECM bioink precursor is provided as an aseptic form. Reconstitution protocol is provided to prepare dECM bioink solution. The final bioink is used for bioprinting self-supporting 3D structures.
Packaging
Each 1EA unit contains 100MG
Legal Information
Product of T&R Biofab
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Three-dimensional (3D) cell printing systems allow the controlled and precise deposition of multiple cells in 3D constructs. Hydrogel materials have been used extensively as printable bioinks owing to their ability to safely encapsulate living cells. However, hydrogel-based bioinks have drawbacks
3D cell-printing technique has been under spotlight as an appealing biofabrication platform due to its ability to precisely pattern living cells in pre-defined spatial locations. In skin tissue engineering, a major remaining challenge is to seek for a suitable source
Trends in biotechnology, 36(8), 787-805 (2018-04-22)
The extracellular matrix (ECM) comprises a complex milieu of proteins and other growth factors that provide mechanical, biophysical, and biochemical cues to cells. The ECM is organ specific, and its detailed composition varies across organs. Bioinks are material formulations and
The ability to print and pattern all the components that make up a tissue (cells and matrix materials) in three dimensions to generate structures similar to tissues is an exciting prospect of bioprinting. However, the majority of the matrix materials
Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.
Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.