Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

904236

Sigma-Aldrich

Sodium taurodeoxycholate-2,2,4,4-d4 solution

100 μg/mL in methanol, ≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$385.00

$385.00


Available to ship onMay 05, 2025Details


Request more information
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 ML
$385.00

About This Item

Empirical Formula (Hill Notation):
C26D4H40NO6SNa
Molecular Weight:
525.71
UNSPSC Code:
12352106
NACRES:
NA.24

$385.00


Available to ship onMay 05, 2025Details


Request more information
Notify Me

Get notified when this item is ready to ship via email.

isotopic purity

≥98 atom % D

Quality Level

assay

≥98% (CP)

form

liquid

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

−20°C

SMILES string

O=C(NCCS(=O)(O[Na])=O)CC[C@H]([C@@H](CC1)[C@]([C@@H](O)C2)(C)C1C(CC3)C2[C@]4(C)[C@@]3([H])C([2H])([2H])[C@H](O)C([2H])([2H])C4)C

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service