Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

903493

Sigma-Aldrich

Fenoprofen-(3-phenoxy-13C6) sodium salt dihydrate

≥99 atom % 13C, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$537.00

$537.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
$537.00

About This Item

Empirical Formula (Hill Notation):
13C6C9H17O5Na
Molecular Weight:
306.24
UNSPSC Code:
12352005
NACRES:
NA.24

$537.00


Check Cart for Availability

Request more information

isotopic purity

≥99 atom % 13C

Quality Level

assay

≥98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service