Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

903175

Sigma-Aldrich

4-(2-Pyrenyl)phenylacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$241.30

$241.30

List Price$254.00
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
$241.30

About This Item

Empirical Formula (Hill Notation):
C24H16O2
Molecular Weight:
336.38
UNSPSC Code:
12352106
NACRES:
NA.23

$241.30

List Price$254.00
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

form

powder

General description

Linker molecule for conjugation of biomolecules to graphene.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service