Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

902861

Sigma-Aldrich

Methyl 4-bromo-1-methyl-1H-pyrrole-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8BrNO2
CAS Number:
Molecular Weight:
218.05
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

assay

≥95%

form

powder or crystals

mp

61-64 °C

functional group

bromo
ester

storage temp.

2-8°C

SMILES string

COC(=O)c1cc(Br)cn1C

InChI

1S/C7H8BrNO2/c1-9-4-5(8)3-6(9)7(10)11-2/h3-4H,1-2H3

InChI key

XYZFZMSLKMPMGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service